Anti-CD3 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HIT3a]

Supplier: Biolegend
300329 300330
CA300329-BLEA 197.88 CAD
CA300329-BL CA300330-BL
Anti-CD3 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HIT3a]
Antibodies
Pacific Blue™ anti-human CD3 [HIT3a]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

* Pacific Blue™ has a maximum emission of 455 nm when it is excited at 405 nm. Prior to using Pacific Blue™ conjugate for flow cytometric analysis, please verify your flow cytometer's capability of exciting and detecting the fluorochrome.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: HIT3a
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR