Anti-CD3 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: OKT3]

Supplier: Biolegend
317313 317314
CA317313-BLEA 197.88 CAD
CA317313-BL CA317314-BL
Anti-CD3 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: OKT3]
Antibodies
Pacific Blue™ anti-human CD3 [OKT3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR