Anti-CD3 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: UCHT1]

Supplier: Biolegend
300417 300418 300431 300442
CA300417-BLEA 427.67 CAD
CA300417-BL CA300418-BL CA300431-BL CA10757-116
Anti-CD3 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: UCHT1]
Antibodies
Pacific Blue™ anti-human CD3 [UCHT1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 500 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis.
For test size, the suggested use of this reagent for immunofluorescent staining is 5 μl per million cells or 5 μl per 100 μl of whole blood.
For μg sizes, the suggested use of this reagent for immunofluorescent staining is 2.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood.
It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR