Anti-CD25 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: BC96]

Supplier: Biolegend
302620 302627
CA302620-BLEA 422.89 CAD
CA302620-BL CA302627-BL
Anti-CD25 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: BC96]
Antibodies
Pacific Blue™ anti-human CD25 [BC96]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis.
For test size, the suggested use of this reagent for immunofluorescent staining is 5 μl per 106 cells in 100 μl volume. 
For μg sizes, the suggested use of this reagent for immunofluorescent staining is 1.0 μg per 106 cells in 100 μl volume.
It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: BC96
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Baboon;Chimpanzee;Cynomolgus;Pigtailed Macaque;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR