Anti-CD20 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 2H7]

Supplier: Biolegend
302319 302320 302328
CA302319-BLEA 197.88 CAD
CA302319-BL CA302320-BL CA302328-BL
Anti-CD20 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 2H7]
Antibodies
Pacific Blue™ anti-human CD20 [2H7]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis.
For test size, the suggested use of this reagent for immunofluorescent staining is 5 μl per 106 cells in 100 μl volume.
For μg sizes, the suggested use of this reagent for immunofluorescent staining is 0.5 μg per 106 cells in 100 μl volume.
It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Baboon;Capuchin Monkey;Chimpanzee;Cynomolgus;Pigtailed Macaque;Rhesus;Squirrel Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR