Anti-CD1c Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: L161]

Supplier: Biolegend
331507 331508
CA331507-BLEA 225.01 CAD
CA331507-BL CA331508-BL
Anti-CD1c Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: L161]
Antibodies
Pacific Blue™ anti-human CD1c [L161]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD1c
Clonality: Monoclonal
Clone: L161
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR