Anti-CD1a Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HI149]

Supplier: Biolegend
300117 300118 300124
CA300117-BLEA 197.88 CAD
CA300117-BL CA300118-BL CA300124-BL
Anti-CD1a Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HI149]
Antibodies
Pacific Blue™ anti-human CD1a [HI149]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis.
For test size, the suggested use of this reagent for immunofluorescent staining is 5 μl per 106 cells in 100 μl volume.
For μg sizes, the suggested use of this reagent for immunofluorescent staining is <0.5 μg per 106 cells in 100 μl volume.
It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD1a
Clonality: Monoclonal
Clone: HI149
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR