Anti-CD16 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 3G8]

Supplier: Biolegend
302021 302024 302032
CA302021-BLEA 427.67 CAD
CA302021-BL CA302024-BL CA302032-BL
Anti-CD16 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 3G8]
Antibodies
Pacific Blue™ anti-human CD16 [3G8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Marmoset, Pigtailed Macaque, Rhesus, Sooty Mangabey, Squirrel Monkey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis.
For test size, the suggested use of this reagent for immunofluorescent staining is 5 μl per 106 cells in 100 μl volume.
For μg sizes, the suggested use of this reagent for immunofluorescent staining is 1.0 μg per 106 cells in 100 μl volume.
It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: 3G8
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Capuchin Monkey;Chimpanzee;Cynomolgus;Marmoset;Pigtailed Macaque;Rhesus;Sooty Mangabey;Squirrel Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR