Anti-CD14 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HCD14]

Supplier: Biolegend
325615 325616
CA11006-564EA 178.73 CAD
CA11006-564 CA325616-BL
Anti-CD14 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HCD14]
Antibodies
Pacific Blue™ anti-human CD14 [HCD14]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: HCD14
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR