Anti-CD14 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: M5E2]

Supplier: Biolegend
301815 301816 301828
CA301815-BLEA 427.67 CAD
CA301815-BL CA301816-BL CA301828-BL
Anti-CD14 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: M5E2]
Antibodies
Pacific Blue™ anti-human CD14 [M5E2]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Capuchin Monkey, Cattle (Bovine, Cow), Chimpanzee, Common Marmoset, Cotton-topped Tamarin, Cynomolgus, Dog (Canine), Rhesus, Pigtailed Macaque, Squirrel Monkey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis.
For test size, the suggested use of this reagent for immunofluorescent staining is 5 μl per million cells or 5 μl per 100 μl of whole blood.
For μg sizes, the suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood.
It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: M5E2
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Capuchin Monkey;Cattle (Bovine;Cow);Chimpanzee;Common Marmoset;Cotton-topped Tamarin;Cynomolgus;Dog (Canine);Rhesus;Pigtailed Macaque;Squirrel Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR