Anti-CD25 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: PC61]

Supplier: Biolegend
102021 102022
CA102021-BLEA 150.01 CAD
CA102021-BL CA102022-BL
Anti-CD25 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: PC61]
Antibodies
Pacific Blue™ anti-mouse CD25 [PC61]; Isotype: Rat IgG1, λ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: PC61
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG1, λ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR