Anti-IgG1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MRG1-58]

Supplier: Biolegend
407405 407406
CA407405-BLEA 132.45 CAD
CA407405-BL CA407406-BL
Anti-IgG1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MRG1-58]
Antibodies
FITC anti-rat IgG1 [MRG1-58]; Isotype: Mouse IgG; Reactivity: Rat; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone: MRG1-58
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR