Anti-g/d TCR Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: UC7-13D5]

Supplier: Biolegend
107503 107504
CA107503-BLEA 142.03 CAD
CA107503-BL CA107504-BL
Anti-g/d TCR Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: UC7-13D5]
Antibodies
FITC anti-mouse TCR γ/δ [UC7-13D5]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: g/d TCR
Clonality: Monoclonal
Clone: UC7-13D5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR