Anti-Ly-6G/Ly-6C (Gr-1) Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RB6-8C5]

Supplier: Biolegend
108405 108406
CA108405-BLEA 38.3 CAD
CA108405-BL CA108406-BL
Anti-Ly-6G/Ly-6C (Gr-1) Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RB6-8C5]
Antibodies
FITC anti-mouse Ly-6G/Ly-6C (Gr-1) [RB6-8C5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ly-6G/Ly-6C (Gr-1)
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR