Anti-IgM Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RMM-1]

Supplier: Biolegend
406505 406506
CA406505-BLEA 132.45 CAD
CA406505-BL CA406506-BL
Anti-IgM Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RMM-1]
Antibodies
FITC anti-mouse IgM [RMM-1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgM
Clonality: Monoclonal
Clone: RMM-1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol IgM
Environmentally Preferable
Antigen synonyms Immunoglobulin M
Antibody type Primary
Clonality Monoclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Clone RMM-1
Reactivity Mouse
Host Rat
Gene ID 16019
Isotype IgG2a kappa
Flow cytometry Yes
Immunogen Mouse Ig cocktail
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
Concentration 0.50 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR