Anti-IgG2b Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RMG2b-1]

Supplier: Biolegend
406705 406706
CA406705-BLEA 132.45 CAD
CA406705-BL CA406706-BL
Anti-IgG2b Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RMG2b-1]
Antibodies
FITC anti-mouse IgG2b [RMG2b-1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgG2b
Clonality: Monoclonal
Clone: RMG2b-1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR