Anti-IgD Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 11-26c.2a]

Supplier: Biolegend
405703 405704
CA405703-BLEA 148.41 CAD
CA405703-BL CA405704-BL
Anti-IgD Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 11-26c.2a]
Antibodies
FITC anti-mouse IgD [11-26c.2a]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgD
Clonality: Monoclonal
Clone: 11-26c.2a
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR