Anti-H-2Kb Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: AF6-88.5]

Supplier: Biolegend
116505 116506
CA116505-BLEA 153.2 CAD
CA116505-BL CA116506-BL
Anti-H-2Kb Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: AF6-88.5]
Antibodies
FITC anti-mouse H-2Kb [AF6-88.5]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse b haplotype, Does not cross-react with other haplotypes (e.g., d, f, j, k, p, q, r, s, u, v); Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kb
Clonality: Monoclonal
Clone: AF6-88.5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse (BALB/c) IgG2a, κ
Reactivity: Mouse b haplotype;Does not cross-react with other haplotypes (e.g.;d;f;j;k;p;q;r;s;u;v)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR