Anti-CD9 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MZ3]

Supplier: Biolegend
124807 124808
CA124807-BLEA 75 CAD
CA124807-BL CA124808-BL
Anti-CD9 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MZ3]
Antibodies
FITC anti-mouse CD9 [MZ3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: MZ3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR