Anti-CD71 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RI7217]

Supplier: Biolegend
113805 113806
CA113805-BLEA 148.41 CAD
CA113805-BL CA113806-BL
Anti-CD71 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RI7217]
Antibodies
FITC anti-mouse CD71 [RI7217]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD71
Clonality: Monoclonal
Clone: RI7217
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR