Anti-CD107a (LAMP-1) Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1D4B]

Supplier: Biolegend
121605 121606
CA121605-BLEA 150.01 CAD
CA121605-BL CA121606-BL
Anti-CD107a (LAMP-1) Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1D4B]
Antibodies
FITC anti-mouse CD107a (LAMP-1) [1D4B]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IF; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD107a (LAMP-1)
Clonality: Monoclonal
Clone: 1D4B
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR