Anti-CD104 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 346-11A]

Supplier: Biolegend
123605 123606
CA123605-BLEA 150.01 CAD
CA123605-BL CA123606-BL
Anti-CD104 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 346-11A]
Antibodies
FITC anti-mouse CD104 [346-11A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD104
Clonality: Monoclonal
Clone: 346-11A
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR