Anti-CD24 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: ML5]

Supplier: Biolegend
311103 311104
CA311103-BLEA 130.86 CAD
CA311103-BL CA311104-BL
Anti-CD24 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: ML5]
Antibodies
FITC anti-human CD24 [ML5]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: ML5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR