Anti-CD19 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HIB19]

Supplier: Biolegend
302205 302206 302256
CA302205-BLEA 79.79 CAD
CA302205-BL CA302206-BL CA10757-714
Anti-CD19 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HIB19]
Antibodies
FITC anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Chimpanzee;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR