Anti-CD18 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TS1/18]
Supplier: Biolegend
302105 302106
CA302105-BLEA 161.18 CAD
CA302105-BL CA302106-BL
Anti-CD18 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TS1/18]
Antibodies
FITC anti-human CD18 [TS1/18]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Swine (Pig, Porcine); Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/More Product Information