Anti-IgM, k Isotype Ctrl Rat Monoclonal Antibody (Biotin) [clone: RTK2118]

Supplier: Biolegend
400803 400804
CA400803-BLEA 132.45 CAD
CA400803-BL CA400804-BL
Anti-IgM, k Isotype Ctrl Rat Monoclonal Antibody (Biotin) [clone: RTK2118]
Antibodies
Biotin Rat IgM, κ Isotype Ctrl [RTK2118]; Isotype: Rat IgM, κ; Reactivity: Rat; Apps: FC, IP, WB; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgM, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK2118
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgM, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR