Anti-IgG3, k Isotype Ctrl Mouse Monoclonal Antibody (Biotin) [clone: MG3-35]

Supplier: Biolegend
401303 401304
CA401303-BLEA 113.3 CAD
CA401303-BL CA401304-BL
Anti-IgG3, k Isotype Ctrl Mouse Monoclonal Antibody (Biotin) [clone: MG3-35]
Antibodies
Biotin Mouse IgG3, κ Isotype Ctrl [MG3-35]; Isotype: Mouse IgG3, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG3, k Isotype Ctrl
Clonality: Monoclonal
Clone: MG3-35
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG3, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR