Anti-IgG1 Mouse Monoclonal Antibody (Biotin) [clone: MRG1-58]

Supplier: Biolegend
407403 407404
CA407403-BLEA 132.45 CAD
CA407403-BL CA407404-BL
Anti-IgG1 Mouse Monoclonal Antibody (Biotin) [clone: MRG1-58]
Antibodies
Biotin anti-rat IgG1 [MRG1-58]; Isotype: Mouse IgG; Reactivity: Rat; Apps: ELISA; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA. For ELISA detection, the suggested use of this reagent is 1.0 μg per ml. It's recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone: MRG1-58
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR