Anti-OX40 Ligand (CD252) Rat Monoclonal Antibody (Biotin) [clone: RM134L]

Supplier: Biolegend
108803 108804
CA108803-BLEA 122.88 CAD
CA108803-BL CA108804-BL
Anti-OX40 Ligand (CD252) Rat Monoclonal Antibody (Biotin) [clone: RM134L]
Antibodies
Biotin anti-mouse CD252 (OX40 Ligand) [RM134L]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: OX40 Ligand (CD252)
Clonality: Monoclonal
Clone: RM134L
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR