Anti-IL-5 Rat Monoclonal Antibody (Biotin) [clone: TRFK4]

Supplier: Biolegend
504401 504402
CA504401-BLEA 161.18 CAD
CA504401-BL CA504402-BL
Anti-IL-5 Rat Monoclonal Antibody (Biotin) [clone: TRFK4]
Antibodies
Biotin anti-mouse IL-5 [TRFK4]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: ELISA Detection, ELISPOT Detection; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 0.25-1.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of mouse IL-5 recombinant protein ranging from 250 to 1 pg/ml are recommended for each ELISA plate. For use as an ELISPOT detection antibody, a concentration range of 1-4 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-5
Clonality: Monoclonal
Clone: TRFK4
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR