Anti-IL-1a Rat Monoclonal Antibody (Biotin) [clone: BL1a-89]

Supplier: Biolegend
512503 512504
CA512503-BLEA 205.86 CAD
CA512503-BL CA512504-BL
Anti-IL-1a Rat Monoclonal Antibody (Biotin) [clone: BL1a-89]
Antibodies
Biotin anti-mouse IL-1α [BL1a-89]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Detection, ELISPOT Detection, WB, IP; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 0.05-0.2 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of mouse IL-1α recobinant protein ranging from 250 to 4 pg/ml are recommended for each ELISA plate. For Western Blotting applications, it is suggested to use 5 μg antibody per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-1a
Clonality: Monoclonal
Clone: BL1a-89
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR