Anti-IL-17A Rat Monoclonal Antibody (Biotin) [clone: TC11-8H4]

Supplier: Biolegend
507001 507002
CA507001-BLEA 188.3 CAD
CA507001-BL CA507002-BL
Anti-IL-17A Rat Monoclonal Antibody (Biotin) [clone: TC11-8H4]
Antibodies
Biotin anti-mouse IL-17A [TC11-8H4]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Detection, ELISPOT Detection; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 0.5-2 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-17 recombinant protein ranging from 4000 to 30 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-17A
Clonality: Monoclonal
Clone:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR