Anti-H-2Kd Mouse Monoclonal Antibody (Biotin) [clone: SF1-1.1]

Supplier: Biolegend
116603 116604
CA116603-BLEA 132.45 CAD
CA116603-BL CA116604-BL
Anti-H-2Kd Mouse Monoclonal Antibody (Biotin) [clone: SF1-1.1]
Antibodies
Biotin anti-mouse H-2Kd [SF1-1.1]; Isotype: Mouse (SJL) IgG2a, κ; Reactivity: Mouse d haplotype, Weakly cross-reacts with H-2k haplotype, Does not cross react with other haplotypes (e.g., b, j, p, q, s, v).; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kd
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR