Anti-CD80 Armenian Hamster Monoclonal Antibody (Biotin) [clone: 16-10A1]

Supplier: Biolegend
104703 104704
CA104703-BLEA 122.88 CAD
CA104703-BL CA104704-BL
Anti-CD80 Armenian Hamster Monoclonal Antibody (Biotin) [clone: 16-10A1]
Antibodies
Biotin anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Cross-Reactivity: Dog (Canine); Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 16-10A1
Conjugation: Biotin
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Cross-Reactivity: Dog (Canine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR