Anti-CD49b (pan-NK cells) Rat Monoclonal Antibody (Biotin) [clone: DX5]

Supplier: Biolegend
108903 108904
CA108903-BLEA 150.01 CAD
CA108903-BL CA108904-BL
Anti-CD49b (pan-NK cells) Rat Monoclonal Antibody (Biotin) [clone: DX5]
Antibodies
Biotin anti-mouse CD49b (pan-NK cells) [DX5]; Isotype: Rat IgM, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD49b (pan-NK cells)
Clonality: Monoclonal
Clone: DX5
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgM, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR