Anti-CD23 Rat Monoclonal Antibody (Biotin) [clone: B3B4]

Supplier: Biolegend
101603 101604
CA101603-BLEA 132.45 CAD
CA101603-BL CA101604-BL
Anti-CD23 Rat Monoclonal Antibody (Biotin) [clone: B3B4]
Antibodies
Biotin anti-mouse CD23 [B3B4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IF; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for other applications.

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: B3B4
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR