Anti-CD2 Rat Monoclonal Antibody (Biotin) [clone: RM2-5]

Supplier: Biolegend
100103 100104
CA100103-BLEA 122.88 CAD
CA100103-BL CA100104-BL
Anti-CD2 Rat Monoclonal Antibody (Biotin) [clone: RM2-5]
Antibodies
Biotin anti-mouse CD2 [RM2-5]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RM2-5
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2b, λ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR