Anti-CD127 (IL-7Ra) Rat Monoclonal Antibody (Biotin) [clone: SB/199]

Supplier: Biolegend
121103 121104
CA121103-BLEA 169.15 CAD
CA121103-BL CA121104-BL
Anti-CD127 (IL-7Ra) Rat Monoclonal Antibody (Biotin) [clone: SB/199]
Antibodies
Biotin anti-mouse CD127 (IL-7Rα) [SB/199]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD127 (IL-7Ra)
Clonality: Monoclonal
Clone: SB/199
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR