Anti-IL-6 Rat Monoclonal Antibody (Biotin) [clone: MQ2-39C3]

Supplier: Biolegend
501201 501202
CA501201-BLEA 161.18 CAD
CA501201-BL CA501202-BL
Anti-IL-6 Rat Monoclonal Antibody (Biotin) [clone: MQ2-39C3]
Antibodies
Biotin anti-human IL-6 [MQ2-39C3]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ELISA Detection, ELISPOT Detection, IF; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA detection applications, a concentration range of 0.5-2.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-6 recombinant protein ranging from 1000 to 8 pg/ml are recommended for each ELISA plate. For use as an ELISPOT detection antibody, a concentration range of 1-4 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-6
Clonality: Monoclonal
Clone: MQ2-39C3
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR