Anti-IL-10 Rat Monoclonal Antibody (Biotin) [clone: JES3-12G8]

Supplier: Biolegend
501501 501502
CA501501-BLEA 161.18 CAD
CA501501-BL CA501502-BL
Anti-IL-10 Rat Monoclonal Antibody (Biotin) [clone: JES3-12G8]
Antibodies
Biotin anti-human IL-10 [JES3-12G8]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ELISA Detection, ELISPOT Detection; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA detection applications, a concentration range of 0.25-1.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-10 recombinant protein ranging from 500 to 2 pg/ml are recommended for each ELISA plate. For use as an ELISPOT detection antibody, a concentration range of 1-4 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-10
Clonality: Monoclonal
Clone: JES3-12G8
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR