Anti-CD4 Mouse Monoclonal Antibody (Biotin) [clone: OKT4]

Supplier: Biolegend
317405 317406
CA317405-BLEA 52.66 CAD
CA317405-BL CA317406-BL
Anti-CD4 Mouse Monoclonal Antibody (Biotin) [clone: OKT4]
Antibodies
Biotin anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: OKT4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR