Anti-CD14 Mouse Monoclonal Antibody (Biotin) [clone: HCD14]

Supplier: Biolegend
325623 325624
CA325623-BLEA 150.01 CAD
CA325623-BL CA325624-BL
Anti-CD14 Mouse Monoclonal Antibody (Biotin) [clone: HCD14]
Antibodies
Biotin anti-human CD14 [HCD14]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, Separation (MojoSort™); Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. For magnetic cell separation with MojoSort™ streptavidin Nanobeads (Cat. No. 480015/480016), the suggested use of this reagent is  20 μg per 10 million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: HCD14
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR