Anti-CD62L Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: DREG-56]

Supplier: Biolegend
304813 304814
CA304813-BLEA 244.16 CAD
CA304813-BL CA304814-BL
Anti-CD62L Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: DREG-56]
Antibodies
APC/Cy7 anti-human CD62L [DREG-56]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cattle (Bovine, Cow); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD62L
Clonality: Monoclonal
Clone: DREG-56
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Cattle (Bovine;Cow)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR