Anti-CD25 Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: BC96]

Supplier: Biolegend
302613 302614
CA302613-BLEA 244.16 CAD
CA302613-BL CA302614-BL
Anti-CD25 Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: BC96]
Antibodies
APC/Cy7 anti-human CD25 [BC96]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: BC96
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Baboon;Chimpanzee;Cynomolgus;Pigtailed Macaque;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR