Anti-IgG Isotype Ctrl Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: HTK888]

Supplier: Biolegend
400911 400912
CA400911-BLEA 151.6 CAD
CA400911-BL CA400912-BL
Anti-IgG Isotype Ctrl Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: HTK888]
Antibodies
APC Armenian Hamster IgG Isotype Ctrl [HTK888]; Isotype: Armenian Hamster IgG; Reactivity: Armenian Hamster; Apps: FC, ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG Isotype Ctrl
Clonality: Monoclonal
Clone: HTK888
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Armenian Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR