Anti-CD8a Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: G28]

Supplier: Biolegend
200609 200610
CA200609-BLEA 151.6 CAD
CA200609-BL CA200610-BL
Anti-CD8a Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: G28]
Antibodies
APC anti-rat CD8a [G28]; Isotype: Mouse IgG2a, κ; Reactivity: Rat; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: G28
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR