Anti-IFNAR-1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MAR1-5A3]

Supplier: Biolegend
127313 127314
CA127313-BLEA 226.6 CAD
CA127313-BL CA127314-BL
Anti-IFNAR-1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MAR1-5A3]
Antibodies
APC anti-mouse IFNAR-1 [MAR1-5A3]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IFNAR-1
Clonality: Monoclonal
Clone: MAR1-5A3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR