Anti-CD45.1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: A20]

Supplier: Biolegend
110713 110714
CA110713-BLEA 70.22 CAD
CA110713-BL CA110714-BL
Anti-CD45.1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: A20]
Antibodies
APC anti-mouse CD45.1 [A20]; Isotype: Mouse (A.SW) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45.1
Clonality: Monoclonal
Clone: A20
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse (A.SW) IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR