Anti-CD25 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: PC61]

Supplier: Biolegend
102011 102012
CA102011-BLEA 86.17 CAD
CA102011-BL CA102012-BL
Anti-CD25 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: PC61]
Antibodies
APC anti-mouse CD25 [PC61]; Isotype: Rat IgG1, λ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: PC61
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG1, λ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR