Anti-CD11c Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: N418]

Supplier: Biolegend
117309 117310
CA117309-BLEA 52.66 CAD
CA117309-BL CA117310-BL
Anti-CD11c Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: N418]
Antibodies
APC anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: N418
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR